Platinum 3% on activated carbon powder wetted with water 22 µm for use as a catalyst, reduced , BASF Catalyst
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Platinium
SP-203-100G
SP-203-25G
SP-203-5G
76124-458EA
1194.8
USD
76124-458
76124-460
76124-462
Platinum 3% on activated carbon powder wetted with water 22 µm for use as a catalyst, reduced , BASF Catalyst
Platinum
3 Percent Platinum on Activated Carbon Powder, Reduced, Water Wet, 22 microns, BASF Catalyst
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 Melting Pt: 1769 Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...