Palladium 5% on activated carbon powder (C-C Coupling) wetted with water 27 µm for use as a catalyst, unreduced , BASF Catalyst
Supplier: Spectrum Chemical Mfg. Corp.
5 Percent Palladium on Activated Carbon Powder, Unreduced, Water Wet (C-C Coupling), 27 microns, BASF Catalyst
Caution: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Formula:
Pd Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...