Premiere Paraffin Dispenser, Newcomer Supply
Supplier: Newcomer Supply
Premiere
6527
76278-806EA 1901.86 USD
76278-806
Premiere Paraffin Dispenser, Newcomer Supply
Paraffin Wax Trimmers and Dispensers
A large capacity tabeletop model with built in temperature control for easy cleaning.
  • Stainless steel basin (2.3 gal.)
  • Heats quickly
  • Easy to use
  • Interior dimensions: 9Ø×10D"
It has a durable industrial metal outer cabinet for long life and built-in temperature controller that heats quickly from room temperature up to 72 °C.
The faucet is heated to help keep the paraffin easily flowing when dispensed.
The lid also has a port in it for inserting a thermometer into the paraffin.
Certifications: The paraffin dispenser is CE certified and the electrical cord is CSA and UL approved.