Airfree® Schlenk Vacuum Traps, with O-Ring Joints, Chemglass
Supplier: Chemglass
AF-0200-01 AF-0200-03 AF-0200-02
80064-378EA 908.33 USD
80064-378 80064-382 80064-380
Airfree® Schlenk Vacuum Traps, with O-Ring Joints, Chemglass
Vacuum Traps
Two-piece vacuum trap with #15 O-Ring joints on both the inlet and outlet ports.
  • Trap body is 57 O.D.×300 mm in length
The seal between the trap stopper and body is made with a 57 mm flat O-Ring flange for total grease-free operation when used with either the double-bank or single-bank manifolds. Trap body is used with a 4300 ml dewar flask.
Delivery information: Complete item consists of the trap stopper and body, one horseshoe clamp, one #331 Viton O-Ring and two #116 Viton O-Rings.