Sulfuric acid 1 N

Supplier: METROHM USA MS

LAB-TI90701
MSPP-TI90701CS 672.92 USD
MSPP-TI90701
Sulfuric acid 1 N
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
MDL Number: MFCD00064589
CAS Number: 7664-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR