L(+)-Arabinose ≥99%, powder

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Pectinose, Pectin Sugar

01654-10KG 01654-5KG 01654-1KG 01654-250G 01654-100G 01654-100G 01654-1KG 01654-250G
102555-622EA 1948.23 USD
102555-622 102561-460 102562-236 102588-578 102616-392 CH01654-100G CH01654-1KG CH01654-250G
L(+)-Arabinose ≥99%, powder
L(+)-Arabinose

L-(+)-Arabinose is used as a one-way or reversible switch for protein expression and also to make arabinosyl nucleosides.

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331
Melting Pt: 154…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00067709
CAS Number: 5328-37-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&vid=6b9738c515009e5720388ea307490836c3b8afa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay ≥99%
Heavy Metals ≤20 ppm
Optical Rotation [a]D20 = +101 to +105°
Loss on Drying ≤1%
Ash ≤0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR