Magnetic Stir Bars, Cowie®
Supplier: Cowie Technology
001.575 001.575-Y 001.575-R 001.525 001.551 001.515 001.522 001.564.8 001.564 001.522-B 001.522-Y 001.538.10 001.525.10 001.513.10 001.551.10 001.525-Y 001.513
76210-946CS 488.36 USD
76210-946 76211-018 76211-020 76211-032 76211-208 76211-212 76211-214 76211-216 76211-220 76211-222 76211-224 76211-226 76211-232 76211-234 76211-236 76211-238 76211-240
Magnetic Stir Bars, Cowie®
Stirrers Magnetic Stirring Bars
Boxed set PTFE stirrer magnets.
  • Provides stable stirring action
  • General-purpose stirrer for a wide variety of applications
Octahedral bars shows similar action to pivot ring type. The dimensions of stirrer bars are nominal and approximate to the stated value by ±5% for length and ±10% for diameter.