Anti-IgG (H+L) Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
2087-05
103264-804EA 238 USD
103264-804
Anti-IgG (H+L) Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat Anti-Human IgG(H+L)-HRP antibody with minimal reactivity to rabbit, mouse, rat, bovine, horse, hamster, goat, sheep, chicken, and guinea pig serum proteins for use in ELISA assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR