Anti-Kappa Goat Antibody

Supplier: Southern Biotechnology
2062-01
100244-352EA 172.62 USD
100244-352
Anti-Kappa Goat Antibody
Antibodies
Purified Goat F(ab')2 Anti-Human Kappa antibody for use in ELISA, flow cytometry, immunocytochemistry, western blotting, and stimulation assays and for immobilization in surface plasmon resonance.

Type: Secondary
Antigen: Kappa
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR