Anti-IgM Goat Antibody (Biotin)

Supplier: Southern Biotechnology
2022-08
100244-230EA 158.23 USD
100244-230
Anti-IgM Goat Antibody (Biotin)
Antibodies
Goat F(ab')2 Anti-Human IgM-Biotin antibody for use in ELISA, flow cytometry, immunocytochemistry, and ELISpot assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR