Anti-IgG1 Goat Antibody (RPE (R-Phycoerythrin))

Supplier: Southern Biotechnology
1070-09S 1070-09
100242-650EA 317.77 USD
100242-650 100242-648
Anti-IgG1 Goat Antibody (RPE (R-Phycoerythrin))
Antibodies
Goat Anti-Mouse IgG1-PE antibody with minimal reactivity to human proteins for use in flow cytometry and multiplex assays.

Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR