Anti-IgG (H+L) Goat Antibody

Supplier: Southern Biotechnology
1031-01
100242-496EA 79.77 USD
100242-496
Anti-IgG (H+L) Goat Antibody
Antibodies
Purified Goat Anti-Mouse IgG(H+L) antibody with minimal reactivity to human proteins for use in ELISA, flow cytometry, immunohistochemistry / immunocytochemistry, western blotting, and ELISpot assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR