Anti-IgM Goat Antibody (APC (Allophycocyanin))

Supplier: Southern Biotechnology
1020-11L 1020-11S
100242-386EA 483.85 USD
100242-386 100242-388
Anti-IgM Goat Antibody (APC (Allophycocyanin))
Antibodies
Goat Anti-Mouse IgM-APC antibody with minimal reactivity to human proteins for use in flow cytometry assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR