Anti-Monocyte/Macrophage Mouse Monoclonal Antibody (Biotin) [clone: KUL01]

Supplier: Southern Biotechnology
8420-08
103265-684EA 439.38 USD
103265-684
Anti-Monocyte/Macrophage Mouse Monoclonal Antibody (Biotin) [clone: KUL01]
Antibodies
The monoclonal antibody KUL01 is useful in the study of the development, distribution, function, and ontogeny of the mononuclear phagocyte system (MPS) of the chicken by exclusively recognizing the cells of the MPS. It identifies chicken monocytes and macrophages as well as interdigitating cells and activated microglia cells. This antibody does not react with B (Bu-1+) or T (CD3+) lymphocytes.

Type: Primary
Antigen: Monocyte/Macrophage
Clonality: Monoclonal
Clone: KUL01
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR