Orthophosphoric acid 0.1 M
Supplier: AQUA SOLUTIONS MS
Synonyms:
Phosphoric acid
6849-20L
101203-700EA
257.28
USD
101203-700
Orthophosphoric acid 0.1 M
Orthophosphoric acid
Formula:
H₃PO₄ MW: 98 g/mol |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 |
Specification Test Results
Molarity | 0.1 M |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...