Glassy carbon, plate, Thickness 3 mm (0.1 in) Type 1

Supplier: Thermo Scientific Chemicals

042821-FF 042821-FI 042821-GH
AA42821-FFEA 140 USD
AA42821-FF AA42821-FI AA42821-GH
Glassy carbon, plate, Thickness 3 mm (0.1 in) Type 1
Glassy carbon
Formula: C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR