Glassy carbon, plate, Thickness 3 mm (0.1 in) Type 2

Supplier: Thermo Scientific Chemicals

042820-FI 042820-FF 042820-GH
AA42820-FIEA 368 USD
AA42820-FI AA42820-FF AA42820-GH
Glassy carbon, plate, Thickness 3 mm (0.1 in) Type 2
Glassy carbon
Formula: C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR