Glassy carbon, plate, Thickness 2 mm (0.08 in) Type 1

Supplier: Thermo Scientific Chemicals

038025-GH 038025-FI
AA38025-GHEA 984 USD
AA38025-GH AA38025-FI
Glassy carbon, plate, Thickness 2 mm (0.08 in) Type 1
Glassy carbon
Formula: C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR