Glassy carbon, plate, Thickness 4 mm (0.16 in) Type 2

Supplier: Thermo Scientific Chemicals

038023-FI 038023-FF 038023-GH
AA38023-FIEA 390 USD
AA38023-FI AA38023-FF AA38023-GH
Glassy carbon, plate, Thickness 4 mm (0.16 in) Type 2
Glassy carbon
Formula: C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR