Klenow, Agilent Technologies
Supplier: Agilent
600069 200410 200410
99902-266EA 156.5 USD
99902-266 CAST200410 ST200410
Klenow, Agilent Technologies
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
The Klenow Fill-In Kit provides the components needed to perform partial fill-in reactions to prepare genomic DNA for insertion into the Lambda FIX II vector.
This kit can also be used to perform a complete fill-in of a 5' overhang to generate blunt ends, which can be directly ligated. Additionally, the kit can be used to radioactively end label a 5' overhang with [alpha-32P]dNTP.
Ordering information: Includes predigested DNA that can be used as a control.
Caution: For research use only. Not for use in diagnostic procedures.