Standard Macintosh Blades, Sklar®
Supplier: Sklar
07-1209
76191-900EA 130.67 USD
76191-900
Standard Macintosh Blades, Sklar®
Scalpels and Scalpel Assemblies Scalpel Blades
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Macintosh Laryngoscope Blades are used for direct, rigid laryngoscopy procedures to view vocal cords, remove foreign objects from the throat, biopsy, or tracheal intubation. This product is a size 3-1/2 American Style Macintosh blade. In general, Macintosh laryngoscope blades are curved. American Style Macintosh blades are slightly shorter than English Style Macintosh blades, and therefore feature less curvature. This blade is not fiber-optic.