Wolff Curette, OR-Grade, Sklar®
Supplier: Sklar
06-4081 06-4082 06-4083 06-4084 06-4085 06-4086
76191-616EA 178.1 USD
76191-616 76191-618 76191-868 76191-870 76191-872 76191-874
Wolff Curette, OR-Grade, Sklar®
Curettes
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
The Wolff Curette is commonly used in dermatological procedures to scrape and remove skin growths and lesions. It may also be known as a Wolff Lupus Curette. It features an oval, cup-like tip with a slight center fenestration.