Humby-Braithwaite Dermatome, OR-Grade, Sklar®
Supplier: Sklar
12-1090
76191-438EA 4450.45 USD
76191-438
Humby-Braithwaite Dermatome, OR-Grade, Sklar®
Scalpels and Scalpel Assemblies Scalpels
Sklar's® Humby-Braithwaite Dermatome is used to collect slices of skin from a donor for performing skin grafts.
  • Latex-free and reusable
Ordering information: The product includes a replaceable blade and an adjustable roller. Blades are available separately.