Diagnostic Vacuum Gauge, Welch
Supplier: Gardner Denver
1526K-10
10836-010EA 1275.31 USD
10836-010
Diagnostic Vacuum Gauge, Welch
Pumps Vacuum Gauges
Diagnostic vacuum gauge, is a rugged, portable vacuum measurement instrument designed specifically for the demands of field use.
  • Strong magnet and kickstand for hands-free use
  • 70 hour battery life, connects easily for accurate readings
  • Self-test mode and field replaceable sensor
  • Can be field calibrated
  • Good/bad sensor (tube) indication
  • Features auto off, on/off soft switch and battery level indication
Highly accurate full range digital millitorr gauge using field-proven thermocouple vacuum sensor technology. This model offers several viewing options including numeric, bar graph, line graph and pump down graph display as well as vacuum analytics for leak, outgassing and pump-down rates. This gauge is capable of displaying in 12 different measurement units (microns, millitorr, torr, mm of Hg, mbar, bar, kPa, Pa, PSIA, inches of Hg, mm and inches of water).
Applications include test vacuum pumps, monitor evacuation and leak test systems and components.
Delivery information: Soft protective boot and hard travel case included.