Test Kits, QAC, LaMotte®
Supplier: LaMotte
7057-01
66170-480EA 152.59 USD
66170-480
Test Kits, QAC, LaMotte®
Environmental Test Kits
Polyelectrolytic titration, ideal for polyquat and also used for low to high concentrations.