QuantiChrom™ Acetylcholinesterase Inhibitor Screening Kit, BioAssay Systems
Supplier: BioAssay Systems
IACE-100
76236-144EA 574.47 USD
76236-144
QuantiChrom™ Acetylcholinesterase Inhibitor Screening Kit, BioAssay Systems
Assays Enzyme Assays
HTS for inhibitor screening and evaluation of acetylcholinesterase inhibitors.
  • Fast and sensitive
  • Linear detection range 10 to 600 U/l for a 10 minute reaction at 37 °C.
  • High-throughput, homogeneous "mix-incubate-measure" type assay
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
  • Method: OD412 nm
  • Samples: acetylcholinesterase
Acetylcholinesterase (EC 3.1.1.7, AChE), also known as RBC cholinesterase, is found primarily in the blood and neural synapses. AChE catalyzes the hydrolysis of the neurotransmitter acetylcholine into choline and acetic acid, a reaction necessary to allow a cholinergic neuron to return to its resting state after activation. Inhibition of the enzyme leads to acetylcholine accumulation, hyperstimulation of nicotinic and muscarinic receptors, and disrupted neurotransmission. AChE inhibition is an important target for the management of Alzheimer's disease. In addition to Alzheimer's disease, AChE inhibitors have been useful in the diagnosis or treatment of diseases such as glaucoma, myasthenia gravis, bladder distention, and more.
QuantiChrom™ acetylcholinesterase inhibitor Assay is based on an improved Ellman method, in which thiocholine produced by the action of acetylcholinesterase forms a yellow color with 5,5'-dithiobis(2-nitrobenzoic acid). The intensity of the product color, measured at 412nm, is proportionate to the enzyme activity in the sample.