Soyabean HiVeg™ Medium, Sterile Powder, HiMedia Laboratories
Supplier: HiMedia
MV011G-5KG MV011G-500G MV011G-2.5KG
95038-662EA 850.4 USD
95038-662 95038-660 89227-186
Soyabean HiVeg™ Medium, Sterile Powder, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Soyabean HiVeg medium, sterile powder is prepared by completely replacing animal-based peptones with vegetable peptones. It is a gamma irradiated sterile powder recommended for the evaluation of sterility in manufacturing process.
The combination of HiVeg hydrolysate and papaic digest of soyabean meal makes this medium nutritious by providing amino acids and long chain peptides for the growth of microorganisms. Dextrose serves as the carbon source and dipotassium phosphate buffers the medium. Sodium chloride maintains the osmotic balance of the medium.