Nitric acid 6.0 N
Supplier: AQUA SOLUTIONS MS
|
Danger
|
Synonyms:
Nitric acid solution
6471-1L
6471-2.5L
6471-500ML
103194-084EA
150.93
USD
103194-084
101202-796
101204-704
Nitric acid 6.0 N
Nitric acid
Formula:
HNO₃ MW: 63.01 g/mol |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 UN: 2031 ADR: 8,II |
Specification Test Results
Normality (N) | 6.0000 ±0.0500 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...