Sodium selenite ≥98%, pale gray powder, Cell Culture Grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
0219474110
0219474190
IC0219474110EA
44.77
USD
IC0219474110
IC0219474190
Sodium selenite ≥98%, pale gray powder, Cell Culture Grade
Sodium selenite
- Grade : Cell Culture Reagent
- Purity : ~98%
- Suitability : Suitable for cell culture
- Product Type : Inorganics
- Chemical Class : Salts
- Vapor Pressure : > 0.001 mm Hg at 20 °C (Lit.)
- Presentation : Pale Gray Powder
- Format : Powder
- NOTES : Selenium Content ≥44.0%
- Solubility : Soluble in water.
Formula:
Na₂SeO₃ Boiling Pt: 684 °C Melting Pt: 217 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00003489 CAS Number: 10102-18-8 |
Specification Test Results
Identity Test | Passes |
Purity |
|
Selenium Content |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...