3,3',5'-Triiodo-L-thyronine sodium salt

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Liothyronine sodium salt, Sodium (S)-2-amino-3-(4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl)propanoate

0219458590 0219458580
76204-140EA 291.34 USD
76204-140 76204-382
3,3',5'-Triiodo-L-thyronine sodium salt
3,3',5'-Triiodo-L-thyronine sodium salt

amino acid found in human plasma and thyroid gland, similar to thyroxine


  • Grade : Cell culture reagent
  • Purity : ≥95%
  • Product Type : Biochemicals
  • Optical Rotation +18 to +22 ° (C=2 IN 1M Hydro-chloric acid : ethanol 1:4 at 25 °C)(Lit.)
  • Presentation : Tan Powder
  • pH : pH (sat'd sol'n): 9 (Lit.)
  • Detection Method : FTIR
  • Solubility : Very slightly soluble in water; Soluble in 1 N HCl : Alcohol mixture (1 : 2); soluble in dilute akilies and 50% Ammonium hydroxide (20 mg/mL - clear, brown solution), 1 N Sodium hydroxide (1 mg/mL); slightly soluble in alcohol, ethanol 96% v/v; practically insoluble in most other organic solvents.

Formula: C₁₅H₁₁I₃NNaO₄
Melting Pt: 205 °C (dec., lit.)
Storage Temperature: Freezer
MDL Number: MFCD00002594
CAS Number: 55-06-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcd85edd53efdbae514ff565b9261d7501305235&vid=bcd85edd53efdbae514ff565b9261d7501305235&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79634f6d30b3d6182d8f3821241fab233da21753&vid=34f453b83c2d311c88350843e083762a5b421efb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity 95%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR