Penicillin G sodium salt, powder, Cell Culture Grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 6-(2-Phenylacetamido)penicillanic acid sodium salt, Benzylpenicillin sodium salt, Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

0219453701 0219453710 0219453725 0219453780
76204-038EA 74.48 USD
76204-038 76204-040 76204-042 76204-044
Penicillin G sodium salt, powder, Cell Culture Grade
Penicillin G sodium salt

Penicillin G also known as benzylpenicillin. It exert a bacteriocidal as well as a bacteriostatic effect on susceptible bacteria by interfering with the final stages of the synthesis of peptidoglycan, a substance in the bacterial cell wall.


  • Grade : Cell culture grade
  • Potency : >1500 u/mg
  • Suitability : Suitable for cell culture
  • Key Applications : Antibiotic
  • Application Areas : Cell biology
  • Product Type : Biochemicals
  • Biochemical Category : Antibiotics
  • Chemical Class : Antibiotics
  • Presentation : Powder
  • Format : Powder
  • Solubility: Soluble in water(H2O):100 mg/mL.
  • Sterilization of Solutions : Solutions should be filter sterilized. Store in small aliquots at appropriate temperature until use.


Storage: protect from light.

Formula: C₁₆H₁₇N₂NaO₄S
MW: 356.38 g/mol
Melting Pt: 255
Storage Temperature: Refrigerator
MDL Number: MFCD00069666
CAS Number: 69-57-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d521a1ac4e4778b46db76d51b25686a922f85c7e&vid=d521a1ac4e4778b46db76d51b25686a922f85c7e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Potency 1500 u/mg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR