D-(+)-Sucrose ≥99%, white crystalline powder for molecular biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: alpha-D-Glucopyranoside, β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, beta-D-Fructofuranosyl alpha-D-glucopyranoside, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, D-(+)-Saccharose, Sucrose, Sugar

0219401890 0219401891 0219401894
76203-956EA 59.22 USD
76203-956 76203-958 76203-960
D-(+)-Sucrose ≥99%, white crystalline powder for molecular biology
D(+)-Sucrose

Sucrose is a disaccharide composed of glucose and fructose.


  • Grade : Molecular biology reagent
  • Purity : >99%
  • Key Applications : Chemical Synthesis; DNA & RNA Purification; Protein extraction; Protein crystallization
  • Application Areas : Molecular biology
  • Product Type : Inorganics
  • Biochemical Category : Biochemicals
  • Vapor Pressure : 0(Lit.)
  • Partition Coefficient : Log P(oct) = -2.66(Lit.)
  • Presentation : White Crystalline Powder:
  • Solubility: Soluble in water (1 g/ 1.3 mL cold water or 0.8 mL hot water), alcohol, acetone, amyl alcohol, pyridine; the aqueous solution is neutral to slightly alkaline.

Formula: C₁₂H₂₂O₁₁
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=435076a509d0a1fe3e39e97cbc0655f624c69377&vid=9c5f000b2aecb14be7ab981d57c20f67808dbbd3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity >99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR