N-Lauroylsarcosine sodium salt ≥95.5%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0219400850 0219400880 0219400883
76204-302EA 47.22 USD
76204-302 76204-304 76204-306
N-Lauroylsarcosine sodium salt ≥95.5%, white powder
N-Lauroylsarcosine sodium salt

N-Lauroylsarcosine is an anionic detergent.


  • Purity : >95.5%
  • Key Applications : Cell Lysis Reagent
  • Electrophoresis : Chromatography Detergents
  • Application Areas : Molecular biology
  • Product Type : Biochemicals
  • Biochemical Category : Biochemicals
  • Extinction Coefficient : (E1%) EM = 3 (280 nm)(Lit.)
  • UV/Visible Absorbance : 220 nm, 265 nm(Lit.)
  • Presentation : White Powder
  • pH : 7.5 - 8.5 (10% aqueous solution)
  • Aggregation Number : 2
  • Critical Micelle Concentration (CMC) : 14.57 mM at 30 °C(Lit.)
  • Solubility : Soluble in water (100 mg/mL - clear to slightly hazy, colorless solution).

Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Flash Pt: 267 °C (512.6 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
UN: 2811
ADR: ,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6ad4befb0a8035d4b0e86f9aecb82da77c3fd0a&vid=e63b123bf0f4167ed5f9105c2f43c9f7ca2865c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity >95.5%
pH (10% aq soln) 7.5 - 8.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR