2-Propanol 99 Molecular_Biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

0219400625 0219400680 0219400690
76204-294EA 42.81 USD
76204-294 76204-296 76204-298
2-Propanol 99 Molecular_Biology
2-Propanol

Isopropyl alcohol is ideal for precipitating nucleic acids. In comparison to ethanol, approximately half the amount is required for precipitation which minimizes the total volume for recovery. Isopropanol is a clear, colorless, polar organic solvent commonly used in chemistry and molecular biology laboratories. It will dissolve a wide range of chemicals, and evaporates quickly.


  • Grade : Molecular Biology Reagent
  • Purity : >99%
  • Key Applications : Precipitating nucleic acids
  • Product Type : Solvents
  • Biochemical Category : Polar Aprotic
  • Auto Ignition : 750 °F
  • Vapor Density : 2.07 (air = 1)
  • Vapor Pressure : 42 hPa at 20 °C
  • Refractive Index : n20/D 1.377(lit.)
  • Presentation : Liquid
  • Water Content : ≤0.2%
  • NOTES : Residue after Evaporation ≤0.001%
  • Solubility : Soluble in water, alcohol, ether,chloroform insoluble in salt.

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.784...0.786 g/cm³ (20 °C)
Flash Pt: 12 °C (53.6 °F) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c77d45f475ffb512ee5843010542e1727d3f93da&vid=fdef7bc521b06b82f56ccae9a79bffd7b9af2d59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity >99%
Solubility in Water Passes
Residue after Evaporation <0.001%
Water <0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR