Agarose, white powder for molecular biology
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0219398325
0219398390
0219398310
0219398350
0219398380
0219398383
76204-290EA
429.32
USD
76204-290
76204-058
76204-288
76204-284
76204-056
76204-286
Agarose, white powder for molecular biology
Agarose
Agarose is a linear polymer consisting of alternating D-galactose and 3,6-anhydro-L-galactose units. Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marinealgae.
- Grade : Molecular Biology reagent
- Suitability : Ideal for nucleic acid electrophoresis, analysis and purification.
- Application Areas : Nucleic Acid Electrophoresis; Nucleic Acid Gel Electrophoresis & Blotting
- Product Type : Biochemicals
- Biochemical Category : Agaroses & Acrylamides
- Chemical Class : Agaroses & Acrylamides
- Presentation : White Powder
- pH : 6 - 7.5 (1% aq soln)
- Electroendosmosis : (-MR) 0.09 - 0.13
- Gel Strength : ≥600 g/cm2 (1.5% gel)
- Gel Temperature : 35 - 37 °C
- Foreign activity : DNase, Rnase and Protease free
- Moisture content : ≤10%
- NOTES : Sulfur ≤0.15%
- Solubility : Soluble in 1.5 g plus 100 mL H2O.
Melting Pt:
87.3...89.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081294 CAS Number: 9012-36-6 |
Specification Test Results
Identity Test | Passes |
Electroendosmosis | 0.09 - 0.13 |
Gel Strength (1.5% gel) |
|
Melting Point (1.5% gel) | 87.3 - 89.5°C |
Moisture |
|
pH (1% aq soln) | 6 - 7.5 |
Sulfur |
|
Gel Temperature | 35 - 37°C |
Dnase / RNase / Protease | Detectable |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...