CleanMax® Clean Manufactured Disposable Coveralls with Elastic Wrists/Ankles, Lakeland Industries
Supplier: Lakeland
CTL417CM-MD CTL417CM-LG CTL417CM-XL CTL417CM-2X CTL417CM-3X CTL417CM-4X CTL417CM-5X
76193-188CS 683.59 USD
76193-188 76193-190 76193-192 76193-194 76193-196 76193-198 76193-200
CleanMax® Clean Manufactured Disposable Coveralls with Elastic Wrists/Ankles, Lakeland Industries
Disposable Garments
Clean manufactured disposable coveralls are individually packaged and folded to promote easier gowning with less excursions.
  • Clean manufactured in silicone-free environment
  • Latex-free
  • Tunneled elastic on wrists with thumb loops
  • Zipper closure
  • For critical environments
  • Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunneled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certifications: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.