Anti-IgG Goat Antibody

Supplier: MP Biomedicals
08641431
IC641431EA 414.75 USD
IC641431
Anti-IgG Goat Antibody
Antibodies
This product is useful for structural and biological studies, as a reference standard, or as starting material for the preparation of an immunogen.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR