Paraplast® X-tra embedding agent for histology
Supplier: Leica Biosystems
Paraplast X-TRA® has been used for tissue embedding.
Made from a unique blend of low molecular weight polymers and paraffin, Paraplast® X-tra has a lower infiltration temperature that eliminates tissue “cooking,” which can cause distortion. Lower viscosity permits complete infiltration, and gives exceptional ribbon continuity with flat sections free of compression distortions. It cuts continuous ribbons of 2μm thickness.
Melting point: 50 to 54°C (122 to 129°F) (ASTM)
Paraplast X-TRA is a registered trademark of Leica Biosystems.
Packaging: Packed in 1kg (2.2lbs.) bags of pellets.
MDL Number:
MFCD00131795 CAS Number: 8002-74-2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...