Paraffin wax blended with synthetic polymers, PureAffin® BLOXX™ preformed paraffin blocks for Peloris® tissue processor
Supplier: CANCER DIAGNOSTICS, INC.
PBX1
10810-016CS 273.98 USD
10810-016
Paraffin wax blended with synthetic polymers, PureAffin® BLOXX™ preformed paraffin blocks for Peloris® tissue processor
Paraffin wax blended with synthetic polymers
Preformed paraffin blocks specially shaped to fit easily into your Peloris®* tissue processor. No measuring or topping off-1 case fills processor chamber. Solid blocks don't spill and create slippery surfaces and no transferring of molten paraffin.
* “The ‘Peloris’ trademark is owned by Vision Biosystems Limited, and CDI does not claim any ownership, affiliation, approval or sponsorship by or from Vision Biosystems Limited with respect to this product.”
CAS Number: 8002-74-2
IVDLogo