Paraffin wax blended with synthetic polymers, Solidification point: 56-58°C, PureAffin® Blue™ for infiltration and embedding
Supplier: CANCER DIAGNOSTICS, INC.
EEPAR2B
10810-018CS
325.52
USD
10810-018
Paraffin wax blended with synthetic polymers, Solidification point: 56-58°C, PureAffin® Blue™ for infiltration and embedding
Paraffin wax blended with synthetic polymers
Pureaffin Blue provides an optimal color contrast to clear and white biopsies. Excellent cutting characteristics. Melting temp 56° C.
- Compatible with all industry paraffins
- Use with open or closed tissue processors
- Blended to provide thorough impregnation of tissues
- Facilitates excellent sectioning characteristics
CAS Number:
8002-74-2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)