Sodium bromide, granular, Lab-Grade
Supplier: AQUA SOLUTIONS MS
S2590-100G
S2590-12KG
S2590-2.5KG
S2590-500G
101223-726EA
32.7
USD
101223-726
101225-150
101226-416
101225-710
Sodium bromide, granular, Lab-Grade
Sodium bromide
Formula:
NaBr MW: 102.89 g/mol |
MDL Number:
MFCD00003475 CAS Number: 7647-15-6 EINECS: 231-599-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...