Brucella HiVeg™ Agar Base Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
MV074-500G MV074-2.5KG MV074-5KG
61000-342EA 256.72 USD
61000-342 89227-372 89227-374
Brucella HiVeg™ Agar Base Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Brucella HiVeg agar base with supplement is recommended for enrichment, isolation and cultivation of Brucella or Campylobacter species from clinical and nonclinical specimens.
Brucella HiVeg agar base is prepared by completely replacing animal based peptones with vegetable peptones, making the medium BSE/TSE risks free. Brucella are intracellular parasites that cause epizootic abortions in animals and septicemic febrile illness or localized infections of bone, tissue or organ systems in humans. Brucella species are highly fastidious and therefore require a nutrient rich medium to be able to grow. Also, Brucella species are highly infective and so extreme care should be taken while handling.