Corn Meal Agar, HiMedia
Supplier: HiMedia
M146-500G M146-2.5KG M146-5KG
61000-028EA 521.87 USD
61000-028 89224-090 89224-104
Corn Meal Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Corn meal agar is recommended for chlamydospore production by C. albicans and the maintenance of fungal stock cultures. Chlamydospore production is an accepted criterion for the identification of Candida species.