Potassium pyroantimonate ≥99.0% for the precipitation of sodium, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Potassium hexahydroxoantimonate, Potassium hexahydroxoantimonate(V), Potassium antimonate
60500-100G
60500-500G
BJ60500-100GEA
155
USD
BJ60500-100G
BJ60500-500G
Potassium pyroantimonate ≥99.0% for the precipitation of sodium, Fluka™
Potassium pyroantimonate
Formula:
KSbO₃·3H₂O MW: 262.9 g/mol Boiling Pt: Dec. (on heating) Storage Temperature: Ambient |
MDL Number:
MFCD00011415 CAS Number: 12208-13-8 EINECS: 235-387-7 UN: 1549 ADR: 6.1 III |
Specification Test Results
Assay (as KSbO₃) | Min. 99 % |
Loss on ignition (600°C, 2 h) | Max. 22 % |
Application test | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...