Potassium nitrate 99.0-100.5%, Puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E252, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Nitrate of Potash
12648-6X1KG
12648-2.5KG
BJ12648-1KGCS
766.05
USD
BJ12648-1KG
BJ12648-2.5KG
BJ-12648-1KG
Potassium nitrate 99.0-100.5%, Puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E252, Fluka™
Potassium nitrate
Formula:
KNO₃ Melting Pt: 335 °C (1.013 hPa) Density: 2.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011409 CAS Number: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III |
Specification Test Results
Assay | 99.0 - 100.5 % |
Assay (calculated to the dried substance) | 99.0 - 100.5 % |
Loss on drying (105°C) | Max. 0.5 % |
pH (5 %, 20°C) | 5-8 |
Ammonium (NH₄) | Max. 50 ppm |
Arsenic (As) | Max. 1 ppm |
Calcium (Ca) | Max. 50 ppm |
Copper (Cu) | Max. 10 ppm |
Iron (Fe) | Max. 10 ppm |
Mercury (Hg) | Max. 1 ppm |
Sodium (Na) | Max. 0.1 % |
Lead (Pb) | Max. 2 ppm |
Zinc (Zn) | Max. 10 ppm |
Heavy metals (as Pb) | Max. 10 ppm |
Chloride (Cl) | Max. 10 ppm |
Nitrite (NO₂) | Max. 5 ppm |
Sulfate (SO₄) | Max. 100 ppm |
Chlorate (ClO₃) | complying |
Appearance of the solution | complying |
Acidity or alkalinity | complying |
Reducible impurities | complying |
Residual solvents | complying |
Identity on K | complying |
Identity on NO₃ | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...