Tantalum disilicide ≥99.5% (Ta basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tantalum silicide

93-7310-5G 93-7310-25G
100199-180EA 108.98 USD
100199-180 100199-178
Tantalum disilicide ≥99.5% (Ta basis)
Tantalum disilicide
Formula: Si₂Ta
Melting Pt: 2200 °C
Density: 9,14
MDL Number: MFCD00049564
CAS Number: 12039-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR