Cerium trichloride hydrate ≥99.9% (Ce basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cerium(III) chloride hydrateCerous chloride

93-5808-500G 93-5808-100G
100205-682EA 96.25 USD
100205-682 100199-638
Cerium trichloride hydrate ≥99.9% (Ce basis)
Cerium trichloride hydrate
Formula: CeCl₃·xH₂O
MW: 246.48 g/mol (anhy)
MDL Number: MFCD00149633
CAS Number: 19423-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR