Molecular sieve A4 (0.4 nm, 4 Å), beads -8+12

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate

93-5406-1000G 93-5406-250G
100199-604EA 155.69 USD
100199-604 100196-738
Molecular sieve A4 (0.4 nm, 4 Å), beads -8+12
Molecular sieve A4 (0.4 nm, 4 Å)

Molecular Sieve

MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR