Molecular sieve A4 (0.4 nm, 4 Å), pellets 1/16''

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate

93-5404-1000G 93-5404-250G
100207-298EA 155.69 USD
100207-298 100210-470
Molecular sieve A4 (0.4 nm, 4 Å), pellets 1/16''
Molecular sieve A4 (0.4 nm, 4 Å)

Molecular Sieve

MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR