, pellets 1/16''

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

93-5402-250G 93-5402-1000G
100196-732EA 62.28 USD
100196-732 100199-598
, pellets 1/16''
Molecular sieve 3A (0.3 nm, 3 Å)

Molecular Sieve

Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR